Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SOX5 SIRNA Assays ›
Gene Symbol:
SOX5
Gene Name:
SRY-box 5
Gene Aliases:
L-SOX5, L-SOX5B, L-SOX5F, LAMSHF
Chromosome Location:
Chr.12: 23529499 - 24562701 on Build GRCh38
UniGene ID:
Hs.657542
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS110097
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261414.1
NP_001248343.1
15
1793
NM_001261415.1
NP_001248344.1
13
1833
NM_006940.4
NP_008871.3
13
1809
NM_152989.3
NP_694534.1
16
2042
NM_178010.2
NP_821078.1
5
609
XM_011520831.2
XP_011519133.1
13
1671
XM_011520832.2
XP_011519134.1
13
1783
XM_011520833.2
XP_011519135.1
13
2112
XM_011520834.2
XP_011519136.1
17
2245
XM_011520835.2
XP_011519137.1
16
2297
XM_011520837.2
XP_011519139.1
13
1854
XM_011520838.2
XP_011519140.1
13
1674
XM_011520842.2
XP_011519144.1
6
734
XM_017019888.1
XP_016875377.1
14
2126
XM_017019889.1
XP_016875378.1
14
2122
XM_017019890.1
XP_016875379.1
17
2261
XM_017019891.1
XP_016875380.1
17
2119
XM_017019892.1
XP_016875381.1
18
2340
XM_017019893.1
XP_016875382.1
15
2200
XM_017019894.1
XP_016875383.1
13
4130
XM_017019895.1
XP_016875384.1
13
1697
XM_017019896.1
XP_016875385.1
17
2258
XM_017019897.1
XP_016875386.1
17
2156
XM_017019898.1
XP_016875387.1
17
2153
XM_017019899.1
XP_016875388.1
16
1865
XM_017019900.1
XP_016875389.1
13
2479
XM_017019901.1
XP_016875390.1
13
6935
GenBank mRNA
AB081588.1
-
16
2040
AB081589.1
-
13
1809
AK074317.1
-
13
1736
AK096569.1
-
13
2103
AK098610.1
-
5
609
AK223536.1
-
5
609
AK299607.1
-
12
1612
AK303970.1
-
13
1833
BC029220.2
-
4
547
BC035227.2
-
4
547
BC047665.2
-
4
547
BC060773.1
-
15
1794
30343519
CR456713.1
-
4
459
JX570584.1
-
14
1854
JX570585.1
-
14
1749
S83308.1
-
4
543
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of chondrocyte differentiation
asymmetric neuroblast division
positive regulation of cartilage development
cellular response to transforming growth factor beta stimulus
positive regulation of mesenchymal stem cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0