Search Thermo Fisher Scientific
Gene Symbol: SRI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256891.1 | NP_001243820.1 | 6, 7 | 563 | ||
NM_001256892.1 | NP_001243821.1 | 6, 7 | 539 | |||
NM_003130.3 | NP_003121.1 | 6, 7 | 563 | |||
NM_198901.1 | NP_944490.1 | 6, 7 | 539 | |||
XM_011516528.1 | XP_011514830.1 | 5, 6 | 433 | |||
GenBank mRNA | AK129770.1 | - | 6, 7 | 532 | ||
AK295225.1 | - | 6, 7 | 396 | |||
AK296601.1 | - | 6, 7 | 522 | |||
BC011025.1 | - | 6, 7 | 521 | 4281626 | ||
BI524104.1 | - | 6, 7 | 524 | |||
BI559592.1 | - | 6, 7 | 557 | |||
BI597920.1 | - | 6, 7 | 559 | |||
BM739144.1 | - | 6, 7 | 539 | |||
BM793972.1 | - | 3, 4 | 281 | |||
BU854353.1 | - | 2, 3 | 202 | |||
L12387.1 | - | 6, 7 | 509 | |||
M32886.1 | - | 6, 7 | 517 |