Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STK3 SIRNA Assays ›
          Gene Symbol:
          STK3
          Gene Name:
          serine/threonine kinase 3
          Gene Aliases:
          KRS1, MST2
          Chromosome Location:
          Chr.8: 98429677 - 98944329 on Build GRCh38
          UniGene ID:
          Hs.492333
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS110314
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STK3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256312.1 NP_001243241.1 5 669
          NM_001256313.1 NP_001243242.1 3 316
          NM_006281.3 NP_006272.2 3 326
          XM_011517248.2 XP_011515550.2 4 2299
          XM_011517251.1 XP_011515553.1 3 340
          XM_011517252.2 XP_011515554.2 4 2299
          XM_011517257.2 XP_011515559.2 4 2299
          XM_017013756.1 XP_016869245.1 4 2299
          XM_017013757.1 XP_016869246.1 4 2299
          XM_017013758.1 XP_016869247.1 3 340
          XM_017013759.1 XP_016869248.1 3 358
          XM_017013760.1 XP_016869249.1 3 311
          XM_017013761.1 XP_016869250.1 3 340
          XM_017013762.1 XP_016869251.1 4 2299
          GenBank mRNA AK075229.1 - 3 312
          AK131363.1 - 5 669
          AK291837.1 - 3 290
          AK307787.1 - 3 316
          AK312660.1 - 3 319
          BC010640.2 - 3 315 3868822
          U26424.1 - 3 323
          U60206.1 - 3 260

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube formation
          endocardium development
          protein phosphorylation
          apoptotic process
          signal transduction
          central nervous system development
          negative regulation of cell proliferation
          signal transduction by protein phosphorylation
          positive regulation of protein binding
          hippo signaling
          intracellular signal transduction
          positive regulation of apoptotic process
          positive regulation of fat cell differentiation
          positive regulation of JNK cascade
          negative regulation of organ growth
          protein stabilization
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of protein kinase B signaling
          primitive hemopoiesis
          cell differentiation involved in embryonic placenta development
          regulation of cell differentiation involved in embryonic placenta development
          positive regulation of protein serine/threonine kinase activity
          negative regulation of canonical Wnt signaling pathway
          hepatocyte apoptotic process
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein binding
          ATP binding
          protein serine/threonine kinase activator activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mtn5c:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0