Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STK3 SIRNA Assays ›
          Gene Symbol:
          STK3
          Gene Name:
          serine/threonine kinase 3
          Gene Aliases:
          KRS1, MST2
          Chromosome Location:
          Chr.8: 98429677 - 98944329 on Build GRCh38
          UniGene ID:
          Hs.492333
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS110316
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STK3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256312.1 NP_001243241.1 7, 8 999
          NM_006281.3 NP_006272.2 5, 6 656
          XM_011517248.2 XP_011515550.2 6, 7 2629
          XM_011517251.1 XP_011515553.1 5, 6 670
          XM_011517253.2 XP_011515555.1 3, 4 339
          XM_011517257.2 XP_011515559.2 6, 7 2629
          XM_017013756.1 XP_016869245.1 6, 7 2629
          XM_017013757.1 XP_016869246.1 6, 7 2629
          XM_017013758.1 XP_016869247.1 5, 6 670
          XM_017013759.1 XP_016869248.1 7, 8 1005
          XM_017013760.1 XP_016869249.1 6, 7 868
          XM_017013761.1 XP_016869250.1 5, 6 670
          XM_017013762.1 XP_016869251.1 6, 7 2629
          GenBank mRNA AK075229.1 - 7, 8 956
          AK131363.1 - 7, 8 999
          AK291837.1 - 5, 6 620
          AK312660.1 - 5, 6 649
          BC010640.2 - 5, 6 645 3868822
          U26424.1 - 5, 6 653
          U60206.1 - 5, 6 590
          Z25422.1 - 2, 3 230

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube formation
          endocardium development
          protein phosphorylation
          apoptotic process
          signal transduction
          central nervous system development
          negative regulation of cell proliferation
          signal transduction by protein phosphorylation
          positive regulation of protein binding
          hippo signaling
          intracellular signal transduction
          positive regulation of apoptotic process
          positive regulation of fat cell differentiation
          positive regulation of JNK cascade
          negative regulation of organ growth
          protein stabilization
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of protein kinase B signaling
          primitive hemopoiesis
          cell differentiation involved in embryonic placenta development
          regulation of cell differentiation involved in embryonic placenta development
          positive regulation of protein serine/threonine kinase activity
          negative regulation of canonical Wnt signaling pathway
          hepatocyte apoptotic process
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein binding
          ATP binding
          protein serine/threonine kinase activator activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qwrcb:80/100.66.75.127:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0