Search
Search

AURKC| Gene Symbol: AURKC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015878.1 | NP_001015878.1 | 6 | 808 | ||
| NM_001015879.1 | NP_001015879.1 | 6 | 667 | |||
| NM_003160.2 | NP_003151.2 | 6 | 672 | |||
| GenBank mRNA | AB017332.1 | - | 6 | 813 | ||
| AF054621.1 | - | 6 | 813 | |||
| AF059681.1 | - | 6 | 868 | |||
| AK301886.1 | - | 5 | 1253 | |||
| AY661554.1 | - | 6 | 667 | |||
| AY714054.1 | - | 6 | 610 | |||
| BC075064.2 | - | 5 | 561 | 30915373 | ||
| EU668345.1 | - | 6 | 868 | |||