Search
Search

SERPINA7| Gene Symbol: SERPINA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000354.5 | NP_000345.2 | 2 | 393 | ||
| XM_005262180.3 | XP_005262237.1 | 2 | 391 | |||
| XM_006724683.2 | XP_006724746.1 | 2 | 391 | |||
| GenBank mRNA | AK310778.1 | - | 2 | 391 | ||
| BC020747.2 | - | 1 | 362 | 4716693 | ||
| BJ996322.1 | - | 2 | 393 | |||
| M14091.1 | - | 1 | 676 | |||