Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF3 SIRNA Assays ›
Gene Symbol:
TCF3
Gene Name:
transcription factor 3
Gene Aliases:
AGM8, E2A, E47, ITF1, TCF-3, VDIR, bHLHb21
Chromosome Location:
Chr.19: 1609290 - 1652590 on Build GRCh38
UniGene ID:
Hs.371282
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS110544
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136139.2
NP_001129611.1
18
1869
NM_003200.3
NP_003191.1
19
1908
XM_006722852.3
XP_006722915.1
19
2083
XM_006722853.3
XP_006722916.1
19
2083
XM_006722855.3
XP_006722918.1
19
2072
XM_006722856.3
XP_006722919.1
19
2073
XM_006722857.3
XP_006722920.1
19
2070
XM_006722858.3
XP_006722921.1
19
2071
XM_011528215.2
XP_011526517.1
18
3381
XM_011528216.2
XP_011526518.1
19
2082
XM_011528217.2
XP_011526519.1
19
2085
XM_011528219.2
XP_011526521.1
19
2080
XM_011528220.2
XP_011526522.1
19
2072
XM_011528221.2
XP_011526523.1
19
2070
XM_011528223.2
XP_011526525.1
19
1991
XM_011528225.2
XP_011526527.1
19
1982
XM_011528226.2
XP_011526528.1
19
1982
XM_011528227.2
XP_011526529.1
17
1831
XM_017027177.1
XP_016882666.1
19
2082
XM_017027178.1
XP_016882667.1
19
2073
XM_017027179.1
XP_016882668.1
19
2070
XM_017027180.1
XP_016882669.1
16
2009
XM_017027181.1
XP_016882670.1
19
1994
XM_017027182.1
XP_016882671.1
19
1984
XM_017027183.1
XP_016882672.1
19
2166
GenBank mRNA
AW028755.1
-
3
328
BC011665.2
-
16
1652
BC110580.1
-
15
1879
40007779
KF303591.1
-
16
1677
M24405.1
-
10
1197
M31222.1
-
1908
M31523.1
-
18
1869
M65214.1
-
3
300
X52078.1
-
15
1623
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
B cell lineage commitment
transcription, DNA-templated
regulation of transcription, DNA-templated
B cell differentiation
immunoglobulin V(D)J recombination
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of muscle cell differentiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
transcription factor binding
mitogen-activated protein kinase kinase kinase binding
enhancer binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
repressing transcription factor binding
vitamin D response element binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-cdcg2:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0