Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TCF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005650.3 | NP_005641.1 | 2 | 4967 | ||
NM_181492.2 | NP_852469.1 | 1 | 4866 | |||
XM_005261722.3 | XP_005261779.1 | 2 | 6170 | |||
XM_006724313.3 | XP_006724376.1 | 2 | 5151 | |||
XM_011530353.2 | XP_011528655.1 | 2 | 5151 | |||
XM_011530354.2 | XP_011528656.1 | 2 | 5151 | |||
GenBank mRNA | AB006630.1 | - | 1 | 4099 | ||
KF851355.1 | - | 2 | 4967 | |||
U19345.1 | - | 1 | 1586 |