Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCP11 SIRNA Assays ›
          Gene Symbol:
          TCP11
          Gene Name:
          t-complex 11
          Gene Aliases:
          D6S230E, FPPR
          Chromosome Location:
          Chr.6: 35118071 - 35141642 on Build GRCh38
          UniGene ID:
          Hs.435371
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS110582
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCP11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001093728.2 NP_001087197.1 10 1583
          NM_001261817.1 NP_001248746.1 10 1568
          NM_001261818.1 NP_001248747.1 9 1445
          NM_001261819.1 NP_001248748.1 9 1430
          NM_001261820.1 NP_001248749.1 11 1789
          NM_001261821.1 NP_001248750.1 11 1774
          NM_018679.5 NP_061149.1 10 1677
          XM_005249339.2 XP_005249396.1 9 1338
          XM_011514830.1 XP_011513132.1 10 1834
          XM_011514831.1 XP_011513133.1 8 1251
          XM_011514832.1 XP_011513134.1 7 1115
          GenBank mRNA AF260330.1 - 7 1126
          AF269223.1 - 9 1361
          AF536532.1 - 11 1629
          AF536533.1 - 10 1616
          AK057234.1 - 10 1400
          AK098590.1 - 8 1172
          AK301778.1 - 10 1459
          AK301975.1 - 9 1366
          AK302022.1 - 9 1331
          AK302375.1 - 10 1508
          AK302456.1 - 10 1489
          AK315078.1 - 10 1610
          AK316403.1 - 10 1392
          AY069943.1 - 9 1385
          BC033729.1 - 10 1529 5167179
          BC040003.1 - 11 1631 5741923
          BC048418.1 - 10 1607 5298037
          BC090050.1 - 11 1635 30387768

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s)

          multicellular organism development
          spermatogenesis
          cell differentiation

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0