Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TSC2 SIRNA Assays ›
          Gene Symbol:
          TSC2
          Gene Name:
          tuberous sclerosis 2
          Gene Aliases:
          LAM, PPP1R160, TSC4
          Chromosome Location:
          Chr.16: 2047804 - 2088720 on Build GRCh38
          UniGene ID:
          Hs.90303
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS111012
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TSC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000548.4 NP_000539.2 6 747
          NM_001077183.2 NP_001070651.1 6 747
          NM_001114382.2 NP_001107854.1 6 747
          NM_001318827.1 NP_001305756.1 5 636
          NM_001318829.1 NP_001305758.1 5 580
          NM_001318832.1 NP_001305761.1 6 648
          XM_005255529.4 XP_005255586.2 6 634
          XM_005255531.4 XP_005255588.2 6 634
          XM_011522636.2 XP_011520938.1 6 634
          XM_011522637.2 XP_011520939.1 6 634
          XM_011522638.2 XP_011520940.2 5 727
          XM_011522639.2 XP_011520941.1 6 634
          XM_011522640.2 XP_011520942.1 6 634
          XM_017023615.1 XP_016879104.1 6 634
          XM_017023616.1 XP_016879105.1 6 634
          XM_017023617.1 XP_016879106.1 5 727
          XM_017023618.1 XP_016879107.1 6 634
          GenBank mRNA AB210000.1 - 6 653
          AK295672.1 - 5 467
          AK295728.1 - 6 648
          BC150300.1 - 6 622 8860381
          BF194994.1 - 3 254
          KJ534954.1 - 5 547
          KJ534955.1 - 5 547
          KJ534958.1 - 5 547
          KJ534982.1 - 5 547
          KJ534984.1 - 5 547
          KJ535039.1 - 5 547
          KJ535091.1 - 5 547
          X75621.1 - 5 565

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          negative regulation of protein kinase activity
          protein import into nucleus
          endocytosis
          cell cycle arrest
          heart development
          protein localization
          negative regulation of cell proliferation
          negative regulation of phosphatidylinositol 3-kinase signaling
          viral process
          vesicle-mediated transport
          regulation of endocytosis
          negative regulation of Wnt signaling pathway
          negative regulation of TOR signaling
          protein kinase B signaling
          positive regulation of GTPase activity
          regulation of insulin receptor signaling pathway
          negative regulation of insulin receptor signaling pathway
          insulin-like growth factor receptor signaling pathway
          positive chemotaxis
          regulation of small GTPase mediated signal transduction
          regulation of cell cycle
          negative regulation of protein kinase B signaling
          GTPase activator activity
          protein binding
          phosphatase binding
          small GTPase binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0