Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VEGFA SIRNA Assays ›
Gene Symbol:
VEGFA
Gene Name:
vascular endothelial growth factor A
Gene Aliases:
MVCD1, VEGF, VPF
Chromosome Location:
Chr.6: 43770209 - 43786487 on Build GRCh38
UniGene ID:
Hs.73793
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS111274
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VEGFA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025366.2
NP_001020537.2
3
1244
NM_001025367.2
NP_001020538.2
3
1244
NM_001025368.2
NP_001020539.2
3
1244
NM_001025369.2
NP_001020540.2
3
1244
NM_001025370.2
NP_001020541.2
3
1244
NM_001033756.2
NP_001028928.1
3
1244
NM_001171622.1
NP_001165093.1
3
1244
NM_001171623.1
NP_001165094.1
3
1244
NM_001171624.1
NP_001165095.1
3
1244
NM_001171625.1
NP_001165096.1
3
1244
NM_001171626.1
NP_001165097.1
3
1244
NM_001171627.1
NP_001165098.1
3
1244
NM_001171628.1
NP_001165099.1
3
1244
NM_001171629.1
NP_001165100.1
3
1244
NM_001171630.1
NP_001165101.1
3
1244
NM_001204384.1
NP_001191313.1
3
1244
NM_001204385.1
NP_001191314.1
3
1244
NM_001287044.1
NP_001273973.1
3
257
NM_001317010.1
NP_001303939.1
3
1244
NM_003376.5
NP_003367.4
3
1244
GenBank mRNA
AB021221.1
-
3
206
AB209485.1
-
1
4386
AB451322.1
-
3
206
AB451451.1
-
3
206
AF022375.1
-
3
907
AF091352.1
-
3
206
AF214570.1
-
3
206
AF430806.1
-
3
206
AF486837.1
-
3
206
AJ010438.1
-
3
206
AK056914.1
-
1
1098
AY047581.1
-
3
222
AY263145.1
-
3
206
AY500353.1
-
3
206
AY766116.1
-
3
206
BC065522.1
-
3
671
BU153227.1
-
3
681
BX442364.2
-
3
249
DQ229900.1
-
3
206
HY066915.1
-
3
257
KT581010.1
-
2
128
KT581011.1
-
3
206
M27281.1
-
3
362
M32977.1
-
3
262
S85192.1
-
3
274
X62568.1
-
3
222
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
ovarian follicle development
patterning of blood vessels
vasculogenesis
response to hypoxia
in utero embryonic development
kidney development
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
cell migration involved in sprouting angiogenesis
positive regulation of neuroblast proliferation
positive regulation of mesenchymal cell proliferation
positive regulation of receptor internalization
platelet degranulation
positive regulation of leukocyte migration
heart morphogenesis
outflow tract morphogenesis
coronary vein morphogenesis
nervous system development
mesoderm development
lactation
positive regulation of cell proliferation
regulation of cell shape
positive regulation of endothelial cell migration
positive regulation of gene expression
monocyte differentiation
macrophage differentiation
lung development
positive regulation of cell migration
regulation of cGMP metabolic process
epithelial cell differentiation
positive regulation of vascular endothelial growth factor receptor signaling pathway
post-embryonic camera-type eye development
positive regulation of protein complex assembly
positive regulation of protein autophosphorylation
activation of protein kinase activity
positive regulation of CREB transcription factor activity
positive regulation of peptidyl-serine phosphorylation
tube formation
endothelial cell chemotaxis
cellular response to vascular endothelial growth factor stimulus
lymph vessel morphogenesis
positive regulation of endothelial cell chemotaxis by VEGF-activated vascular endothelial growth factor receptor signaling pathway
positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway
VEGF-activated neuropilin signaling pathway
growth
eye photoreceptor cell development
positive regulation of tyrosine phosphorylation of Stat3 protein
negative regulation of apoptotic process
surfactant homeostasis
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MAP kinase activity
positive regulation of blood vessel endothelial cell migration
positive regulation of angiogenesis
positive regulation of cell adhesion
positive regulation of transcription from RNA polymerase II promoter
vascular endothelial growth factor receptor signaling pathway
cell maturation
camera-type eye morphogenesis
cardiac muscle fiber development
branching morphogenesis of an epithelial tube
positive regulation of axon extension involved in axon guidance
artery morphogenesis
positive regulation of epithelial cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
positive regulation of positive chemotaxis
induction of positive chemotaxis
positive regulation of cellular component movement
positive regulation of cell division
positive regulation of focal adhesion assembly
primitive erythrocyte differentiation
mammary gland alveolus development
cardiac vascular smooth muscle cell development
coronary artery morphogenesis
regulation of transcription from RNA polymerase II promoter in response to hypoxia
positive regulation of transcription from RNA polymerase II promoter in response to hypoxia
cellular response to hypoxia
dopaminergic neuron differentiation
commissural neuron axon guidance
positive regulation of protein kinase C signaling
positive regulation of cell migration involved in sprouting angiogenesis
positive regulation of branching involved in ureteric bud morphogenesis
regulation of retinal ganglion cell axon guidance
positive regulation of peptidyl-tyrosine autophosphorylation
positive regulation of p38MAPK cascade
positive regulation of histone deacetylase activity
positive regulation of retinal ganglion cell axon guidance
positive regulation of protein kinase D signaling
fibronectin binding
cytokine activity
platelet-derived growth factor receptor binding
vascular endothelial growth factor receptor binding
protein binding
growth factor activity
heparin binding
chemoattractant activity
identical protein binding
protein homodimerization activity
vascular endothelial growth factor receptor 1 binding
vascular endothelial growth factor receptor 2 binding
protein heterodimerization activity
receptor agonist activity
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0