Search
Search

WAS| Gene Symbol: WAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000377.2 | NP_000368.1 | 1 | 168 | ||
| XM_011543977.2 | XP_011542279.1 | 1 | 173 | |||
| XM_017029786.1 | XP_016885275.1 | 1 | 212 | |||
| GenBank mRNA | BC002961.1 | - | 1 | 147 | ||
| BC012738.2 | - | 1 | 139 | 3688197 | ||
| BI910072.1 | - | 1 | 184 | |||
| U12707.1 | - | 1 | 145 | |||
| U19927.1 | - | 1 | 135 | |||