Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT7A SIRNA Assays ›
          Gene Symbol:
          WNT7A
          Gene Name:
          Wnt family member 7A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 13818108 - 13880121 on Build GRCh38
          UniGene ID:
          Hs.72290
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS111362
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNT7A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004625.3 NP_004616.2 2 521
          XM_011534091.2 XP_011532393.1 3 268
          GenBank mRNA BC008811.2 - 2 521 3949579
          D83175.1 - 2 246
          U53476.1 - 2 240

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001100473.
          This siRNA also targets the following transcripts in Mouse: NM_009527, AK004683, BC057586, M89801, BC049093.

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          cartilage condensation
          angiogenesis
          chondrocyte differentiation
          neurotransmitter secretion
          axonogenesis
          synapse assembly
          sex differentiation
          asymmetric protein localization
          dorsal/ventral pattern formation
          positive regulation of endothelial cell migration
          skeletal muscle satellite cell activation
          skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
          Wnt signaling pathway
          cerebellar granule cell differentiation
          cell proliferation in forebrain
          central nervous system vasculogenesis
          establishment of cell polarity
          regulation of axon diameter
          response to estradiol
          somatic stem cell population maintenance
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          non-canonical Wnt signaling pathway
          Wnt signaling pathway involved in wound healing, spreading of epidermal cells
          synaptic vesicle recycling
          embryonic digit morphogenesis
          negative regulation of apoptotic process
          response to estrogen
          cell fate commitment
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          somatic stem cell division
          stem cell development
          negative regulation of neurogenesis
          positive regulation of synapse assembly
          palate development
          positive regulation of epithelial cell proliferation involved in wound healing
          oviduct development
          canonical Wnt signaling pathway
          dendritic spine morphogenesis
          uterus morphogenesis
          lens fiber cell development
          cellular response to transforming growth factor beta stimulus
          positive regulation of canonical Wnt signaling pathway
          positive regulation of protein localization to synapse
          excitatory synapse assembly
          positive regulation of excitatory synapse assembly
          positive regulation of excitatory postsynaptic potential
          receptor binding
          frizzled binding
          cytokine activity
          protein binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lcrps:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0