Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
生物製劑委託開發製造與臨床試驗服務
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF41 SIRNA Assays ›
Gene Symbol:
ZNF41
Gene Name:
zinc finger protein 41
Gene Aliases:
MRX89
Chromosome Location:
Chr.X: 47444691 - 47483671 on Build GRCh38
UniGene ID:
Hs.496074
Species:
Human
siRNA ID
HSS111538
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF41
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324139.1
NP_001311068.1
4
808
NM_001324140.1
NP_001311069.1
5
1098
NM_001324141.1
NP_001311070.1
4
837
NM_001324142.1
NP_001311071.1
5
1114
NM_001324143.1
NP_001311072.1
4
1150
NM_001324144.1
NP_001311073.1
5
1188
NM_001324145.1
NP_001311074.1
4
853
NM_001324147.1
NP_001311076.1
5
1159
NM_001324148.1
NP_001311077.1
5
1204
NM_001324149.1
NP_001311078.1
4
877
NM_001324150.1
NP_001311079.1
6
1286
NM_001324151.1
NP_001311080.1
5
1228
NM_001324152.1
NP_001311081.1
5
1274
NM_001324153.1
NP_001311082.1
5
1138
NM_001324154.1
NP_001311083.1
4
1025
NM_001324155.1
NP_001311084.1
4
1049
NM_001324156.1
NP_001311085.1
4
830
NM_001324157.1
NP_001311086.1
4
815
NM_007130.3
NP_009061.1
5
1501
NM_153380.3
NP_700359.1
5
1374
XM_006724550.3
XP_006724613.1
5
1227
XM_006724555.3
XP_006724618.1
5
1522
XM_017029810.1
XP_016885299.1
5
1602
XM_017029811.1
XP_016885300.1
5
1663
XM_017029812.1
XP_016885301.1
5
1584
XM_017029813.1
XP_016885302.1
5
1133
XM_017029814.1
XP_016885303.1
5
1573
XM_017029815.1
XP_016885304.1
5
1622
XM_017029816.1
XP_016885305.1
5
1061
XM_017029817.1
XP_016885306.1
5
1021
GenBank mRNA
AB209520.1
-
1
1210
AK290021.1
-
5
1155
AK294858.1
-
4
750
BC015023.1
-
5
1384
3908304
X60155.2
-
5
1140
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-fwnls:80/10.239.167.40:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0