Search Thermo Fisher Scientific
Gene Symbol: CSDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007553.2 | NP_001007554.1 | 11, 12 | 1708 | ||
NM_001130523.2 | NP_001123995.1 | 11, 12 | 1762 | |||
NM_001242891.1 | NP_001229820.1 | 12, 13 | 1855 | |||
NM_001242892.1 | NP_001229821.1 | 10, 11 | 1321 | |||
NM_001242893.1 | NP_001229822.1 | 9, 10 | 1228 | |||
NM_007158.5 | NP_009089.4 | 10, 11 | 1615 | |||
GenBank mRNA | AB020692.1 | - | 11, 12 | 1622 | ||
AF070542.1 | - | 9, 10 | 1212 | |||
AF077054.1 | - | 10, 11 | 1531 | |||
AK022516.1 | - | 9, 10 | 1130 | |||
AK022528.1 | - | 9, 10 | 1130 | |||
AK290146.1 | - | 10, 11 | 1253 | |||
AY049788.1 | - | 8, 9 | 1117 | |||
BC032446.1 | - | 10, 11 | 1550 | 5111213 | ||
BX648561.1 | - | 10, 11 | 1452 | |||
BX648780.1 | - | 10, 11 | 1549 | |||
CR749431.1 | - | 11, 12 | 1694 |