Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BAG6 SIRNA Assays ›
          Gene Symbol:
          BAG6
          Gene Name:
          BCL2 associated athanogene 6
          Gene Aliases:
          BAG-6, BAT3, D6S52E, G3
          Chromosome Location:
          Chr.6: 31639028 - 31660900 on Build GRCh38
          UniGene ID:
          Hs.440900
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS111846
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BAG6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098534.1 NP_001092004.1 18, 19 2703
          NM_001199697.1 NP_001186626.1 18, 19 2455
          NM_001199698.1 NP_001186627.1 18, 19 2833
          NM_004639.3 NP_004630.3 18, 19 2875
          NM_080702.2 NP_542433.1 18, 19 2833
          NM_080703.2 NP_542434.1 18, 19 2857
          XM_011514892.2 XP_011513194.1 19, 20 3957
          XM_017011275.1 XP_016866764.1 19, 20 3984
          XM_017011276.1 XP_016866765.1 19, 20 2974
          XM_017011277.1 XP_016866766.1 19, 20 2834
          XM_017011278.1 XP_016866767.1 19, 20 2909
          XM_017011279.1 XP_016866768.1 19, 20 3304
          XM_017011280.1 XP_016866769.1 19, 20 2876
          XM_017011281.1 XP_016866770.1 19, 20 3038
          XM_017011282.1 XP_016866771.1 19, 20 2830
          XM_017011283.1 XP_016866772.1 19, 20 3981
          XM_017011284.1 XP_016866773.1 19, 20 3951
          XM_017011285.1 XP_016866774.1 18, 19 2774
          XM_017011286.1 XP_016866775.1 18, 19 2793
          XM_017011287.1 XP_016866776.1 19, 20 3924
          XM_017011288.1 XP_016866777.1 18, 19 2711
          XM_017011289.1 XP_016866778.1 18, 19 3843
          XM_017011290.1 XP_016866779.1 19, 20 3984
          XM_017011291.1 XP_016866780.1 19, 20 3984
          XM_017011292.1 XP_016866781.1 18, 19 3813
          XM_017011293.1 XP_016866782.1 17, 18 2606
          XM_017011294.1 XP_016866783.1 17, 18 3738
          XM_017011295.1 XP_016866784.1 19, 20 3984
          XM_017011296.1 XP_016866785.1 18, 19 3813
          XM_017011297.1 XP_016866786.1 18, 19 2711
          XM_017011298.1 XP_016866787.1 18, 19 3813
          GenBank mRNA AK302695.1 - 18, 19 2406
          BC003133.1 - 18, 19 2751 2957896
          M33519.1 - 18, 19 2810

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          immune system process
          ubiquitin-dependent protein catabolic process
          transport
          synaptonemal complex assembly
          spermatogenesis
          brain development
          embryo development
          covalent chromatin modification
          internal peptidyl-lysine acetylation
          cell differentiation
          lung development
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          negative regulation of apoptotic process
          negative regulation of proteolysis
          protein stabilization
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          tail-anchored membrane protein insertion into ER membrane
          positive regulation of ERAD pathway
          maintenance of unfolded protein involved in ERAD pathway
          protein localization to cytosolic proteasome complex involved in ERAD pathway
          protein binding
          Hsp70 protein binding
          polyubiquitin binding
          ubiquitin protein ligase binding
          ribosome binding
          misfolded protein binding
          proteasome binding
          ubiquitin-specific protease binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0