Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NELFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002904.5 | NP_002895.3 | 8 | 1015 | ||
XM_006715205.3 | XP_006715268.1 | 8 | 884 | |||
XM_011514913.2 | XP_011513215.1 | 8 | 955 | |||
XM_017011299.1 | XP_016866788.1 | 8 | 940 | |||
GenBank mRNA | AK300717.1 | - | 8 | 798 | ||
AK300898.1 | - | 8 | 890 | |||
AK302652.1 | - | 8 | 1002 | |||
AK303152.1 | - | 7 | 1062 | |||
BC025235.1 | - | 8 | 866 | 4876045 | ||
BC050617.2 | - | 8 | 882 | 6168937 | ||
JQ313904.1 | - | 7 | 811 | |||
L03411.1 | - | 8 | 897 | |||
X16105.1 | - | 5 | 681 |