Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CUL3 SIRNA Assays ›
          Gene Symbol:
          CUL3
          Gene Name:
          cullin 3
          Gene Aliases:
          CUL-3, PHA2E
          Chromosome Location:
          Chr.2: 224470150 - 224585397 on Build GRCh38
          UniGene ID:
          Hs.372286
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS112303
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CUL3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257197.1 NP_001244126.1 7 1348
          NM_001257198.1 NP_001244127.1 8 1229
          NM_003590.4 NP_003581.1 8 1546
          XM_006712800.2 XP_006712863.2 8 1219
          XM_011511994.2 XP_011510296.1 7 1103
          XM_011511995.1 XP_011510297.1 8 1131
          XM_011511996.2 XP_011510298.1 8 1195
          GenBank mRNA AB014517.2 - 8 1474
          AF062537.1 - 8 1247
          AF064087.1 - 8 1239
          AK291151.1 - 8 1196
          AK299644.1 - 7 958
          AK308496.1 - 8 1229
          AY337761.1 - 8 1692
          BC039598.1 - 8 1491 5784147
          BC092409.1 - 8 1491 6168545
          U58089.1 - 4 585

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106923, XM_008767226, XM_008767227, XM_008767228, XM_008767229, and others.
          This siRNA also targets the following transcripts in Mouse: NM_016716, BC027304, AK129174, AF129738.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          protein polyubiquitination
          trophectodermal cellular morphogenesis
          protein monoubiquitination
          ER to Golgi vesicle-mediated transport
          cell cycle arrest
          mitotic metaphase plate congression
          integrin-mediated signaling pathway
          gastrulation
          negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
          positive regulation of cell proliferation
          Wnt signaling pathway
          cell migration
          protein ubiquitination
          stem cell division
          cell projection organization
          positive regulation of cytokinesis
          negative regulation of Rho protein signal transduction
          embryonic cleavage
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          stress fiber assembly
          proteasome-mediated ubiquitin-dependent protein catabolic process
          fibroblast apoptotic process
          positive regulation of mitotic metaphase/anaphase transition
          COPII vesicle coating
          liver morphogenesis
          negative regulation of canonical Wnt signaling pathway
          intrinsic apoptotic signaling pathway
          ubiquitin-protein transferase activity
          protein binding
          cyclin binding
          POZ domain binding
          ubiquitin protein ligase binding
          protein homodimerization activity
          protein heterodimerization activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0