Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CUL3 SIRNA Assays ›
          Gene Symbol:
          CUL3
          Gene Name:
          cullin 3
          Gene Aliases:
          CUL-3, PHA2E
          Chromosome Location:
          Chr.2: 224470150 - 224585397 on Build GRCh38
          UniGene ID:
          Hs.372286
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS112305
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CUL3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257197.1 NP_001244126.1 15 2477
          NM_001257198.1 NP_001244127.1 16 2358
          NM_003590.4 NP_003581.1 16 2675
          XM_006712800.2 XP_006712863.2 16 2348
          XM_011511994.2 XP_011510296.1 15 2232
          XM_011511995.1 XP_011510297.1 16 2260
          XM_011511996.2 XP_011510298.1 16 2324
          GenBank mRNA AB014517.2 - 16 2603
          AF052147.1 - 9 1098
          AF062537.1 - 16 2376
          AF064087.1 - 16 2368
          AK098531.1 - 2 2043
          AK291151.1 - 16 2325
          AK299644.1 - 15 2087
          AK308496.1 - 16 2358
          AY337761.1 - 16 2821
          BC031844.1 - 9 1103
          BC039598.1 - 16 2620 5784147
          BC092409.1 - 16 2620 6168545
          U58089.1 - 12 1714

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_016716, BC027304, AK003885, AK129174, AF129738.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          protein polyubiquitination
          trophectodermal cellular morphogenesis
          protein monoubiquitination
          ER to Golgi vesicle-mediated transport
          cell cycle arrest
          mitotic metaphase plate congression
          integrin-mediated signaling pathway
          gastrulation
          negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
          positive regulation of cell proliferation
          Wnt signaling pathway
          cell migration
          protein ubiquitination
          stem cell division
          cell projection organization
          positive regulation of cytokinesis
          negative regulation of Rho protein signal transduction
          embryonic cleavage
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          stress fiber assembly
          proteasome-mediated ubiquitin-dependent protein catabolic process
          fibroblast apoptotic process
          positive regulation of mitotic metaphase/anaphase transition
          COPII vesicle coating
          liver morphogenesis
          negative regulation of canonical Wnt signaling pathway
          intrinsic apoptotic signaling pathway
          ubiquitin-protein transferase activity
          protein binding
          cyclin binding
          POZ domain binding
          ubiquitin protein ligase binding
          protein homodimerization activity
          protein heterodimerization activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0