Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA2 SIRNA Assays ›
Gene Symbol:
PPFIA2
Gene Name:
PTPRF interacting protein alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 81257975 - 81759748 on Build GRCh38
UniGene ID:
Hs.506216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS112393
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220473.2
NP_001207402.1
9
1372
NM_001220474.2
NP_001207403.1
8
1318
NM_001220475.2
NP_001207404.1
9
1184
NM_001220476.2
NP_001207405.1
9
1184
NM_001220477.2
NP_001207406.1
7
1191
NM_001220478.2
NP_001207407.1
6
1116
NM_001282536.1
NP_001269465.1
7
879
NM_003625.4
NP_003616.2
10
1235
XM_006719670.3
XP_006719733.1
7
8427
XM_011538906.2
XP_011537208.1
8
1312
XM_011538907.2
XP_011537209.1
8
1312
XM_017020079.1
XP_016875568.1
10
1219
XM_017020080.1
XP_016875569.1
9
1366
XM_017020081.1
XP_016875570.1
9
1366
XM_017020082.1
XP_016875571.1
10
1219
XM_017020083.1
XP_016875572.1
9
1366
XM_017020084.1
XP_016875573.1
9
1366
XM_017020085.1
XP_016875574.1
9
1366
XM_017020086.1
XP_016875575.1
10
1219
XM_017020087.1
XP_016875576.1
9
1353
XM_017020088.1
XP_016875577.1
9
1599
XM_017020089.1
XP_016875578.1
8
1312
XM_017020090.1
XP_016875579.1
10
1219
XM_017020091.1
XP_016875580.1
9
1366
XM_017020092.1
XP_016875581.1
9
1366
XM_017020093.1
XP_016875582.1
9
1366
XM_017020094.1
XP_016875583.1
9
1144
XM_017020095.1
XP_016875584.1
9
1366
XM_017020096.1
XP_016875585.1
8
1291
XM_017020097.1
XP_016875586.1
10
1219
XM_017020098.1
XP_016875587.1
10
1219
XM_017020099.1
XP_016875588.1
8
1312
XM_017020100.1
XP_016875589.1
9
1366
XM_017020101.1
XP_016875590.1
10
1219
XM_017020102.1
XP_016875591.1
8
1278
XM_017020103.1
XP_016875592.1
9
1366
XM_017020104.1
XP_016875593.1
9
1366
XM_017020105.1
XP_016875594.1
8
1291
XM_017020106.1
XP_016875595.1
9
1366
XM_017020107.1
XP_016875596.1
10
1219
XM_017020108.1
XP_016875597.1
10
1219
XM_017020109.1
XP_016875598.1
9
1366
XM_017020110.1
XP_016875599.1
8
1524
XM_017020111.1
XP_016875600.1
9
1366
XM_017020112.1
XP_016875601.1
7
1191
XM_017020113.1
XP_016875602.1
7
1191
XM_017020114.1
XP_016875603.1
7
1191
XM_017020115.1
XP_016875604.1
7
1191
XM_017020116.1
XP_016875605.1
7
1191
XM_017020117.1
XP_016875606.1
6
926
XM_017020118.1
XP_016875607.1
8
1166
XM_017020119.1
XP_016875608.1
7
1191
XM_017020120.1
XP_016875609.1
7
879
GenBank mRNA
AB210009.1
-
9
1370
AF034799.1
-
10
1243
AK126971.1
-
8
1278
AK294505.1
-
6
926
AK297372.1
-
8
1166
AK302658.1
-
7
850
BC104912.1
-
9
1168
8143915
BC143485.1
-
9
1168
9051993
DA329958.1
-
2
212
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
neurotransmitter secretion
glutamate secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline