Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA2 SIRNA Assays ›
Gene Symbol:
PPFIA2
Gene Name:
PTPRF interacting protein alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 81257975 - 81759748 on Build GRCh38
UniGene ID:
Hs.506216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS112394
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220473.2
NP_001207402.1
4, 5
706
NM_001220474.2
NP_001207403.1
3, 4
652
NM_001220475.2
NP_001207404.1
4, 5
518
NM_001220476.2
NP_001207405.1
4, 5
518
NM_001220477.2
NP_001207406.1
2, 3
525
NM_001220478.2
NP_001207407.1
2, 3
525
NM_001282536.1
NP_001269465.1
2, 3
213
NM_003625.4
NP_003616.2
5, 6
569
XM_006719670.3
XP_006719733.1
2, 3
7761
XM_011538906.2
XP_011537208.1
3, 4
646
XM_011538907.2
XP_011537209.1
3, 4
646
XM_017020079.1
XP_016875568.1
5, 6
553
XM_017020080.1
XP_016875569.1
4, 5
700
XM_017020081.1
XP_016875570.1
4, 5
700
XM_017020082.1
XP_016875571.1
5, 6
553
XM_017020083.1
XP_016875572.1
4, 5
700
XM_017020084.1
XP_016875573.1
4, 5
700
XM_017020085.1
XP_016875574.1
4, 5
700
XM_017020086.1
XP_016875575.1
5, 6
553
XM_017020087.1
XP_016875576.1
4, 5
687
XM_017020088.1
XP_016875577.1
4, 5
933
XM_017020089.1
XP_016875578.1
3, 4
646
XM_017020090.1
XP_016875579.1
5, 6
553
XM_017020091.1
XP_016875580.1
4, 5
700
XM_017020092.1
XP_016875581.1
4, 5
700
XM_017020093.1
XP_016875582.1
4, 5
700
XM_017020094.1
XP_016875583.1
5, 6
553
XM_017020095.1
XP_016875584.1
4, 5
700
XM_017020096.1
XP_016875585.1
4, 5
700
XM_017020097.1
XP_016875586.1
5, 6
553
XM_017020098.1
XP_016875587.1
5, 6
553
XM_017020099.1
XP_016875588.1
3, 4
646
XM_017020100.1
XP_016875589.1
4, 5
700
XM_017020101.1
XP_016875590.1
5, 6
553
XM_017020102.1
XP_016875591.1
4, 5
687
XM_017020103.1
XP_016875592.1
4, 5
700
XM_017020104.1
XP_016875593.1
4, 5
700
XM_017020105.1
XP_016875594.1
4, 5
700
XM_017020106.1
XP_016875595.1
4, 5
700
XM_017020107.1
XP_016875596.1
5, 6
553
XM_017020108.1
XP_016875597.1
5, 6
553
XM_017020109.1
XP_016875598.1
4, 5
700
XM_017020110.1
XP_016875599.1
4, 5
933
XM_017020111.1
XP_016875600.1
4, 5
700
XM_017020112.1
XP_016875601.1
2, 3
525
XM_017020113.1
XP_016875602.1
2, 3
525
XM_017020114.1
XP_016875603.1
2, 3
525
XM_017020115.1
XP_016875604.1
2, 3
525
XM_017020116.1
XP_016875605.1
2, 3
525
XM_017020117.1
XP_016875606.1
2, 3
335
XM_017020118.1
XP_016875607.1
3, 4
500
XM_017020119.1
XP_016875608.1
2, 3
525
XM_017020120.1
XP_016875609.1
2, 3
213
GenBank mRNA
AB210009.1
-
4, 5
704
AF034799.1
-
5, 6
577
AK126971.1
-
3, 4
612
AK294505.1
-
2, 3
335
AK297372.1
-
3, 4
500
AK299853.1
-
2, 3
474
AK302658.1
-
2, 3
184
AK307524.1
-
5, 6
553
BC104912.1
-
4, 5
502
8143915
BC143485.1
-
4, 5
502
9051993
BQ027545.1
-
2, 3
365
DB083201.1
-
2, 3
213
DC321269.1
-
2, 3
494
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
neurotransmitter secretion
glutamate secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline