Search
Search

DNAH17| Gene Symbol: DNAH17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173628.3 | NP_775899.3 | 65, 66 | 10660 | ||
| XM_011525416.2 | XP_011523718.1 | 65, 66 | 10696 | |||
| XM_017025261.1 | XP_016880750.1 | 23, 24 | 3920 | |||
| GenBank mRNA | AB290170.1 | - | 8, 9 | 1361 | ||
| AJ000522.1 | - | 4, 5 | 687 | |||
| AL122077.1 | - | 6, 7 | 983 | |||
| AL832652.1 | - | 10, 11 | 1731 | |||