Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other B3GALNT1 SIRNA Assays ›
Gene Symbol:
B3GALNT1
Gene Name:
beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group)
Gene Aliases:
B3GALT3, GLCT3, GLOB, Gb4Cer, P, P1, beta3Gal-T3, galT3
Chromosome Location:
Chr.3: 161083883 - 161105528 on Build GRCh38
UniGene ID:
Hs.418062
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS112783
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: B3GALNT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001038628.1
NP_001033717.1
8
914
NM_003781.3
NP_003772.1
5
591
NM_033167.2
NP_149357.1
6
710
NM_033168.2
NP_149358.1
4
496
NM_033169.2
NP_149359.1
5
564
XM_005247859.4
XP_005247916.1
6
654
XM_005247861.3
XP_005247918.1
7
774
XM_005247862.3
XP_005247919.1
6
632
XM_005247864.3
XP_005247921.1
5
589
XM_006713803.3
XP_006713866.1
7
773
XM_006713804.3
XP_006713867.1
4
714
XM_006713805.3
XP_006713868.1
5
833
XM_017007402.1
XP_016862891.1
6
728
XM_017007403.1
XP_016862892.1
6
676
XM_017007404.1
XP_016862893.1
6
684
XM_017007405.1
XP_016862894.1
5
557
XM_017007406.1
XP_016862895.1
7
751
XM_017007407.1
XP_016862896.1
7
900
XM_017007408.1
XP_016862897.1
6
803
XM_017007409.1
XP_016862898.1
5
696
XM_017007410.1
XP_016862899.1
7
783
XM_017007411.1
XP_016862900.1
5
705
XM_017007412.1
XP_016862901.1
4
630
GenBank mRNA
AB050855.1
-
1
253
AB050856.1
-
5
507
AF132731.1
-
2
220
AF154848.1
-
5
540
AK022491.1
-
5
543
AK095917.1
-
4
496
AK223439.1
-
5
504
AK295552.1
-
2
208
AY359049.1
-
4
466
BC028571.1
-
8
836
4838965
BC047618.1
-
5
486
5259171
CR457170.1
-
1
145
DB172381.1
-
2
288
Y15062.1
-
1
145
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
oligosaccharide biosynthetic process
UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity
galactosylgalactosylglucosylceramide beta-D-acetylgalactosaminyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0