Search
Search

EED| Gene Symbol: EED | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308007.1 | NP_001294936.1 | 9 | 1797 | ||
| NM_003797.4 | NP_003788.2 | 9 | 1797 | |||
| XM_011545330.1 | XP_011543632.1 | 9 | 1637 | |||
| XM_011545331.1 | XP_011543633.1 | 9 | 1637 | |||
| XM_017018512.1 | XP_016874001.1 | 9 | 1637 | |||
| GenBank mRNA | AA811323.1 | - | 4 | 600 | ||
| AF070418.1 | - | 9 | 1179 | |||
| AF078933.1 | - | 9 | 943 | |||
| AF080227.1 | - | 9 | 1295 | |||
| AF099032.1 | - | 9 | 1041 | |||
| AK292120.1 | - | 9 | 1406 | |||
| AK301265.1 | - | 9 | 913 | |||
| BC047672.2 | - | 9 | 910 | |||
| BC068995.1 | - | 9 | 958 | 4637702 | ||
| U90651.1 | - | 9 | 1041 | |||