Search
Search

RNGTT| Gene Symbol: RNGTT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286426.1 | NP_001273355.1 | 7 | 984 | ||
| NM_001286428.1 | NP_001273357.1 | 6 | 874 | |||
| NM_003800.4 | NP_003791.3 | 7 | 984 | |||
| XM_017011401.1 | XP_016866890.1 | 7 | 1071 | |||
| XM_017011402.1 | XP_016866891.1 | 7 | 1071 | |||
| GenBank mRNA | AB009022.1 | - | 7 | 764 | ||
| AB009023.1 | - | 7 | 764 | |||
| AB009024.1 | - | 7 | 764 | |||
| AB012142.1 | - | 7 | 919 | |||
| AB012143.1 | - | 7 | 919 | |||
| AF025654.1 | - | 7 | 1052 | |||
| AK295719.1 | - | 8 | 862 | |||
| AK299777.1 | - | 6 | 864 | |||
| AK312407.1 | - | 7 | 955 | |||
| BC019954.1 | - | 7 | 951 | 3923156 | ||