Search
Search

FBP2| Gene Symbol: FBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003837.3 | NP_003828.2 | 2 | 291 | ||
| GenBank mRNA | BC113632.1 | - | 2 | 221 | 8322684 | |
| BC117477.1 | - | 2 | 221 | 40126028 | ||
| BM686555.1 | - | 2 | 299 | |||
| CR536483.1 | - | 2 | 185 | |||
| Y10812.2 | - | 2 | 252 | |||