Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TRIM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003852.3 | NP_003843.3 | 9 | 1558 | ||
NM_015905.2 | NP_056989.2 | 9 | 1558 | |||
GenBank mRNA | AF009353.1 | - | 9 | 1331 | ||
AF119042.1 | - | 9 | 1459 | |||
AK075306.1 | - | 9 | 1558 | |||
AK302679.1 | - | 10 | 1431 | |||
AK312610.1 | - | 9 | 1686 | |||
BC028689.2 | - | 9 | 1456 | 4827559 |