Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SYNJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160302.1 | NP_001153774.1 | 32 | 4278 | ||
NM_001160306.1 | NP_001153778.1 | 28 | 4161 | |||
NM_003895.3 | NP_003886.3 | 32 | 4294 | |||
NM_203446.2 | NP_982271.2 | 33 | 4318 | |||
XM_017028494.1 | XP_016883983.1 | 31 | 4236 | |||
XM_017028495.1 | XP_016883984.1 | 31 | 4294 | |||
XM_017028496.1 | XP_016883985.1 | 30 | 4197 | |||
XM_017028497.1 | XP_016883986.1 | 31 | 4334 | |||
XM_017028498.1 | XP_016883987.1 | 29 | 4158 | |||
XM_017028499.1 | XP_016883988.1 | 28 | 4168 | |||
XM_017028500.1 | XP_016883989.1 | 33 | 4308 | |||
XM_017028501.1 | XP_016883990.1 | 33 | 4305 | |||
XM_017028502.1 | XP_016883991.1 | 32 | 4257 | |||
XM_017028503.1 | XP_016883992.1 | 31 | 4221 | |||
XM_017028504.1 | XP_016883993.1 | 30 | 4182 | |||
XM_017028505.1 | XP_016883994.1 | 29 | 4192 | |||
GenBank mRNA | AB020717.2 | - | 31 | 4220 | ||
AF009039.1 | - | 33 | 4250 | |||
AF009040.1 | - | 32 | 4226 | |||
AK295706.1 | - | 10 | 1259 | |||
AK304044.1 | - | 7 | 1141 | |||
BC098395.1 | - | 28 | 4128 | |||
BC136603.1 | - | 28 | 4087 | 9020593 | ||
DQ421853.1 | - | 6 | 993 |