Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF7 SIRNA Assays ›
Gene Symbol:
ARHGEF7
Gene Name:
Rho guanine nucleotide exchange factor 7
Gene Aliases:
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
Chromosome Location:
Chr.13: 111114619 - 111305734 on Build GRCh38
UniGene ID:
Hs.508738
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS113107
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113511.2
NP_001106983.1
11, 12
2190
NM_001113512.2
NP_001106984.1
9, 10
2040
NM_001113513.1
NP_001106985.1
9, 10
1132
NM_001320851.1
NP_001307780.1
8, 9
913
NM_001320852.1
NP_001307781.1
10, 11
2127
NM_001320853.1
NP_001307782.1
9, 10
1038
NM_001320854.1
NP_001307783.1
5, 6
652
NM_003899.3
NP_003890.1
9, 10
1245
NM_145735.3
NP_663788.1
10, 11
2127
XM_006719956.3
XP_006720019.1
10, 11
1255
XM_011521133.2
XP_011519435.1
10, 11
1255
XM_017020814.1
XP_016876303.1
10, 11
1255
XM_017020815.1
XP_016876304.1
8, 9
1248
XM_017020816.1
XP_016876305.1
9, 10
1023
XM_017020817.1
XP_016876306.1
8, 9
1248
XM_017020818.1
XP_016876307.1
10, 11
1123
XM_017020819.1
XP_016876308.1
9, 10
1132
XM_017020820.1
XP_016876309.1
9, 10
1255
XM_017020821.1
XP_016876310.1
9, 10
950
XM_017020822.1
XP_016876311.1
8, 9
946
XM_017020823.1
XP_016876312.1
8, 9
1723
XM_017020824.1
XP_016876313.1
10, 11
1123
XM_017020825.1
XP_016876314.1
8, 9
874
XM_017020826.1
XP_016876315.1
10, 11
1123
XM_017020827.1
XP_016876316.1
8, 9
947
XM_017020828.1
XP_016876317.1
6, 7
571
XM_017020829.1
XP_016876318.1
6, 7
571
GenBank mRNA
AB177848.1
-
10, 11
2128
AB177849.1
-
7, 8
851
AJ844630.1
-
2, 3
169
AK295472.1
-
5, 6
577
AK300272.1
-
9, 10
1025
AK308897.1
-
5, 6
652
AL834228.1
-
10, 11
1311
BC033905.1
-
9, 10
1237
5262912
BC050521.1
-
9, 10
1104
BC060776.1
-
11, 12
1532
5271977
D63476.1
-
9, 10
1221
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
Golgi organization
signal transduction
nervous system development
positive regulation of fibroblast migration
lamellipodium assembly
positive regulation of protein binding
regulation of Rho protein signal transduction
intracellular signal transduction
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of apoptotic process
positive regulation of GTPase activity
ephrin receptor signaling pathway
focal adhesion assembly
regulation of small GTPase mediated signal transduction
positive regulation of growth hormone secretion
positive regulation of substrate adhesion-dependent cell spreading
regulation of GTP binding
positive regulation of lamellipodium morphogenesis
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0