Search
Search

WASL| Gene Symbol: WASL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003941.3 | NP_003932.3 | 9 | 1580 | ||
| GenBank mRNA | AK289795.1 | - | 9 | 1493 | ||
| AM295156.1 | - | 9 | 1243 | |||
| BC052955.1 | - | 9 | 1530 | 5264663 | ||
| CR749438.1 | - | 10 | 1485 | |||
| D88460.1 | - | 9 | 1497 | |||