Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: USP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304284.1 | NP_001291213.1 | 22 | 3578 | ||
NM_004505.3 | NP_004496.2 | 14 | 3047 | |||
XM_011524050.1 | XP_011522352.1 | 18 | 3796 | |||
XM_011524051.2 | XP_011522353.1 | 20 | 2891 | |||
XM_011524052.2 | XP_011522354.1 | 19 | 3005 | |||
XM_011524053.2 | XP_011522355.1 | 20 | 2918 | |||
XM_011524054.2 | XP_011522356.1 | 20 | 2885 | |||
XM_011524055.2 | XP_011522357.1 | 18 | 2812 | |||
XM_011524056.2 | XP_011522358.1 | 15 | 2765 | |||
XM_011524057.2 | XP_011522359.1 | 18 | 2928 | |||
XM_011524058.2 | XP_011522360.1 | 19 | 2994 | |||
XM_011524059.2 | XP_011522361.1 | 19 | 2994 | |||
XM_017025290.1 | XP_016880779.1 | 17 | 3634 | |||
GenBank mRNA | AB449915.1 | - | 13 | 1348 | ||
AY143550.1 | - | 14 | 3047 | |||
BX647719.1 | - | 22 | 3579 | |||
X63546.1 | - | 14 | 3044 | |||
X63547.1 | - | 15 | 3255 |