Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: COG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018714.2 | NP_061184.1 | 6 | 1300 | ||
GenBank mRNA | AB037802.1 | - | 6 | 1262 | ||
AK025633.1 | - | 3 | 426 | |||
AK098224.1 | - | 6 | 1020 | |||
BC021985.2 | - | 6 | 1265 | 2821581 |