Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: STOML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256672.1 | NP_001243601.1 | 3 | 485 | ||
NM_001256675.1 | NP_001243604.1 | 3 | 485 | |||
NM_001256676.1 | NP_001243605.1 | 3 | 299 | |||
NM_001256677.1 | NP_001243606.1 | 4 | 616 | |||
NM_001324226.1 | NP_001311155.1 | 3 | 299 | |||
NM_001324227.1 | NP_001311156.1 | 3 | 299 | |||
NM_001324228.1 | NP_001311157.1 | 3 | 363 | |||
NM_001324229.1 | NP_001311158.1 | 3 | 485 | |||
NM_001324230.1 | NP_001311159.1 | 4 | 616 | |||
NM_004809.4 | NP_004800.2 | 3 | 485 | |||
XM_005254792.4 | XP_005254849.1 | 3 | 1389 | |||
XM_006720773.3 | XP_006720836.1 | 3 | 1390 | |||
GenBank mRNA | AF074953.1 | - | 3 | 347 | ||
AF156557.1 | - | 3 | 493 | |||
AK075244.1 | - | 3 | 381 | |||
AK091531.1 | - | 3 | 285 | |||
AK222665.1 | - | 3 | 368 | |||
AK300799.1 | - | 4 | 500 | |||
AL109665.1 | - | 3 | 339 | |||
AL533460.3 | - | 3 | 276 | |||
BC034379.1 | - | 3 | 431 | 5185908 | ||
BC065249.1 | - | 3 | 431 | 6059393 | ||
BX464941.2 | - | 3 | 299 | |||
CR542089.1 | - | 3 | 360 | |||
DQ064605.1 | - | 3 | 360 | |||
Y16522.1 | - | 3 | 404 |