Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCAR1 SIRNA Assays ›
          Gene Symbol:
          BCAR1
          Gene Name:
          BCAR1, Cas family scaffolding protein
          Gene Aliases:
          CAS, CAS1, CASS1, CRKAS, P130Cas
          Chromosome Location:
          Chr.16: 75229030 - 75268053 on Build GRCh38
          UniGene ID:
          Hs.479747
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS114271
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCAR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001170714.1 NP_001164185.1 3 450
          NM_001170715.1 NP_001164186.1 2 155
          NM_001170716.1 NP_001164187.1 2 112
          NM_001170717.1 NP_001164188.1 2 174
          NM_001170718.1 NP_001164189.1 2 199
          NM_001170719.1 NP_001164190.1 2 289
          NM_001170720.1 NP_001164191.1 3 304
          NM_014567.3 NP_055382.2 2 174
          XM_005256258.4 XP_005256315.1 2 374
          XM_005256259.4 XP_005256316.1 2 3286
          XM_005256260.4 XP_005256317.1 2 295
          XM_011523475.2 XP_011521777.1 2 323
          XM_017023897.1 XP_016879386.1 2 881
          XM_017023898.1 XP_016879387.1 2 881
          XM_017023899.1 XP_016879388.1 2 881
          GenBank mRNA AB040024.1 - 2 169
          AF218451.1 - 2 152
          AJ242987.2 - 2 149
          AK027608.1 - 2 199
          AK124526.1 - 2 1594
          AK124815.1 - 2 158
          AK293808.1 - 3 450
          AK294513.1 - 2 205
          AK295809.1 - 2 84
          AK302617.1 - 3 304
          AK314722.1 - 2 174
          AY545071.1 - 1 28
          BC062556.1 - 1 29 6427808
          DA418642.1 - 2 174
          DC310094.1 - 2 289
          DC317947.1 - 2 115

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          actin filament organization
          cell adhesion
          epidermal growth factor receptor signaling pathway
          G-protein coupled receptor signaling pathway
          integrin-mediated signaling pathway
          cell proliferation
          insulin receptor signaling pathway
          positive regulation of endothelial cell migration
          cell migration
          positive regulation of cell migration
          cellular response to hepatocyte growth factor stimulus
          regulation of apoptotic process
          platelet-derived growth factor receptor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          neurotrophin TRK receptor signaling pathway
          hepatocyte growth factor receptor signaling pathway
          antigen receptor-mediated signaling pathway
          T cell receptor signaling pathway
          B cell receptor signaling pathway
          cell division
          cell chemotaxis
          signal transducer activity
          protein binding
          SH3 domain binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0