Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCAR1 SIRNA Assays ›
          Gene Symbol:
          BCAR1
          Gene Name:
          BCAR1, Cas family scaffolding protein
          Gene Aliases:
          CAS, CAS1, CASS1, CRKAS, P130Cas
          Chromosome Location:
          Chr.16: 75229030 - 75268053 on Build GRCh38
          UniGene ID:
          Hs.479747
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS114273
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCAR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001170714.1 NP_001164185.1 3 608
          NM_001170715.1 NP_001164186.1 2 313
          NM_001170716.1 NP_001164187.1 2 270
          NM_001170717.1 NP_001164188.1 2 332
          NM_001170718.1 NP_001164189.1 2 357
          NM_001170719.1 NP_001164190.1 2 447
          NM_014567.3 NP_055382.2 2 332
          XM_005256258.4 XP_005256315.1 2 532
          XM_005256259.4 XP_005256316.1 2 3444
          XM_005256260.4 XP_005256317.1 2 453
          XM_011523475.2 XP_011521777.1 2 481
          XM_017023897.1 XP_016879386.1 2 1039
          XM_017023898.1 XP_016879387.1 2 1039
          XM_017023899.1 XP_016879388.1 2 1039
          GenBank mRNA AB040024.1 - 2 327
          AF218451.1 - 2 310
          AJ242987.2 - 2 307
          AK124526.1 - 2 1752
          AK124815.1 - 2 316
          AK293808.1 - 3 608
          AK294513.1 - 2 363
          AK295809.1 - 2 242
          AK314722.1 - 2 332
          AY545071.1 - 1 186
          BC062556.1 - 1 187 6427808
          DA418642.1 - 2 332
          DC310094.1 - 2 447
          DC317947.1 - 2 273

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          actin filament organization
          cell adhesion
          epidermal growth factor receptor signaling pathway
          G-protein coupled receptor signaling pathway
          integrin-mediated signaling pathway
          cell proliferation
          insulin receptor signaling pathway
          positive regulation of endothelial cell migration
          cell migration
          positive regulation of cell migration
          cellular response to hepatocyte growth factor stimulus
          regulation of apoptotic process
          platelet-derived growth factor receptor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          neurotrophin TRK receptor signaling pathway
          hepatocyte growth factor receptor signaling pathway
          antigen receptor-mediated signaling pathway
          T cell receptor signaling pathway
          B cell receptor signaling pathway
          cell division
          cell chemotaxis
          signal transducer activity
          protein binding
          SH3 domain binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0