Search
Search

MTFR1| Gene Symbol: MTFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145838.1 | NP_001139310.1 | 4 | 679 | ||
| NM_001145839.1 | NP_001139311.1 | 6 | 910 | |||
| NM_014637.3 | NP_055452.3 | 6 | 910 | |||
| XM_006716484.2 | XP_006716547.2 | 6 | 882 | |||
| XM_011517626.1 | XP_011515928.1 | 6 | 864 | |||
| XM_011517627.2 | XP_011515929.1 | 6 | 800 | |||
| XM_011517628.2 | XP_011515930.1 | 5 | 1017 | |||
| GenBank mRNA | AK298945.1 | - | 5 | 408 | ||
| AK304716.1 | - | 6 | 824 | |||
| AK316202.1 | - | 5 | 450 | |||
| BC036116.1 | - | 6 | 824 | 5299040 | ||
| BC043498.2 | - | 6 | 809 | 6057524 | ||
| BX537854.1 | - | 4 | 571 | |||
| BX647507.1 | - | 6 | 829 | |||
| CR456910.1 | - | 4 | 641 | |||
| D13634.1 | - | 5 | 658 | |||