Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTSS1 SIRNA Assays ›
Gene Symbol:
MTSS1
Gene Name:
metastasis suppressor 1
Gene Aliases:
MIM, MIMA, MIMB
Chromosome Location:
Chr.8: 124550770 - 124728507 on Build GRCh38
UniGene ID:
Hs.336994
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS114749
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTSS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282971.1
NP_001269900.1
14, 15
2131
NM_001282974.1
NP_001269903.1
13, 14
2044
NM_014751.5
NP_055566.3
13, 14
2119
XM_005251111.1
XP_005251168.1
13, 14
2424
XM_005251113.1
XP_005251170.1
12, 13
2229
XM_005251118.1
XP_005251175.1
13, 14
1902
XM_006716700.1
XP_006716763.1
13, 14
2421
XM_006716701.1
XP_006716764.1
12, 13
2412
XM_006716702.1
XP_006716765.1
13, 14
2373
XM_006716703.1
XP_006716766.1
12, 13
2178
XM_006716704.1
XP_006716767.1
14, 15
2094
XM_006716705.1
XP_006716768.1
14, 15
2046
XM_006716706.1
XP_006716769.1
13, 14
1851
XM_011517403.1
XP_011515705.1
12, 13
1878
XM_011517404.2
XP_011515706.1
11, 12
13590
XM_011517407.1
XP_011515709.1
4, 5
1078
XM_017014086.1
XP_016869575.1
12, 13
2409
XM_017014087.1
XP_016869576.1
13, 14
2349
XM_017014088.1
XP_016869577.1
12, 13
2337
XM_017014089.1
XP_016869578.1
12, 13
2334
XM_017014090.1
XP_016869579.1
11, 12
2970
XM_017014091.1
XP_016869580.1
13, 14
1858
XM_017014092.1
XP_016869581.1
11, 12
2217
XM_017014093.1
XP_016869582.1
13, 14
2082
XM_017014094.1
XP_016869583.1
13, 14
2034
XM_017014095.1
XP_016869584.1
14, 15
2022
XM_017014096.1
XP_016869585.1
14, 15
2019
XM_017014097.1
XP_016869586.1
13, 14
2007
XM_017014098.1
XP_016869587.1
12, 13
1890
XM_017014099.1
XP_016869588.1
12, 13
1887
XM_017014100.1
XP_016869589.1
12, 13
1839
XM_017014101.1
XP_016869590.1
11, 12
1669
GenBank mRNA
AB007889.2
-
10, 11
1301
AF086645.1
-
13, 14
2030
AK027015.1
-
14, 15
1897
AK126651.1
-
1, 2
831
AK126989.1
-
6, 7
875
AK295681.1
-
6, 7
930
AK302036.1
-
7, 8
1375
AK310792.1
-
5, 6
841
BC023998.1
-
7, 8
891
4826982
EF560715.1
-
13, 14
1675
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
movement of cell or subcellular component
plasma membrane organization
cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
microspike assembly
actin cytoskeleton organization
negative regulation of epithelial cell proliferation
renal tubule morphogenesis
cellular response to fluid shear stress
glomerulus morphogenesis
nephron tubule epithelial cell differentiation
xenophagy
epithelial cell proliferation involved in renal tubule morphogenesis
actin monomer binding
receptor binding
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0