Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI1 SIRNA Assays ›
Gene Symbol:
ABI1
Gene Name:
abl interactor 1
Gene Aliases:
ABI-1, ABLBP4, E3B1, NAP1BP, SSH3BP, SSH3BP1
Chromosome Location:
Chr.10: 26746596 - 26861087 on Build GRCh38
UniGene ID:
Hs.508148
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS115190
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012750.2
NP_001012768.1
5, 6
807
NM_001012751.2
NP_001012769.1
5, 6
807
NM_001012752.2
NP_001012770.1
4, 5
792
NM_001178116.1
NP_001171587.1
6, 7
858
NM_001178119.1
NP_001171590.1
5, 6
807
NM_001178120.1
NP_001171591.1
5, 6
807
NM_001178121.1
NP_001171592.1
4, 5
792
NM_001178122.1
NP_001171593.1
5, 6
807
NM_001178123.1
NP_001171594.1
5, 6
807
NM_001178124.1
NP_001171595.1
4, 5
792
NM_001178125.1
NP_001171596.1
3, 4
615
NM_005470.3
NP_005461.2
5, 6
807
XM_005252332.3
XP_005252389.1
4, 5
607
XM_005252333.3
XP_005252390.1
4, 5
607
XM_006717359.2
XP_006717422.1
5, 6
706
XM_006717360.2
XP_006717423.1
5, 6
706
XM_006717361.2
XP_006717424.1
4, 5
607
XM_011519289.2
XP_011517591.1
5, 6
706
XM_017015458.1
XP_016870947.1
5, 6
706
XM_017015459.1
XP_016870948.1
5, 6
706
GenBank mRNA
AB040151.1
-
5, 6
750
AB209268.1
-
5, 6
658
AF001628.1
-
5, 6
631
AF006516.1
-
5, 6
649
AF260262.1
-
4, 5
691
AF540955.1
-
4, 5
580
AK126803.1
-
6, 7
757
AK291823.1
-
4, 5
653
AK296751.1
-
3, 4
357
AK298646.1
-
3, 4
512
BC013238.1
-
5, 6
655
BC024254.2
-
4, 5
643
3531592
FJ380057.1
-
5, 6
583
FJ380058.1
-
5, 6
583
FJ380059.1
-
4, 5
568
U87166.1
-
5, 6
664
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
somitogenesis
movement of cell or subcellular component
transmembrane receptor protein tyrosine kinase signaling pathway
actin polymerization or depolymerization
negative regulation of cell proliferation
viral process
peptidyl-tyrosine phosphorylation
megakaryocyte development
Fc-gamma receptor signaling pathway involved in phagocytosis
vascular endothelial growth factor receptor signaling pathway
positive regulation of protein tyrosine kinase activity
lamellipodium morphogenesis
cell-cell adhesion
protein binding
cytoskeletal protein binding
protein tyrosine kinase activator activity
protein complex binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-gxlt5:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0