Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AP1M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300887.1 | NP_001287816.1 | 10 | 1156 | ||
NM_005498.4 | NP_005489.2 | 10 | 1150 | |||
GenBank mRNA | AF020797.2 | - | 10 | 1113 | ||
AK023863.1 | - | 10 | 1151 | |||
AK222946.1 | - | 10 | 1116 | |||
AK315689.1 | - | 10 | 1141 | |||
AW370781.1 | - | 3 | 517 | |||
BC003387.2 | - | 10 | 1088 | 3446133 | ||
BC003612.2 | - | 10 | 1087 | 2988458 | ||
BC005021.1 | - | 10 | 1156 | 3139433 |