Search
Search

STAM2| Gene Symbol: STAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005843.5 | NP_005834.4 | 10 | 1256 | ||
| GenBank mRNA | AF042273.1 | - | 10 | 1015 | ||
| AF042274.1 | - | 10 | 1015 | |||
| AK292265.1 | - | 10 | 951 | |||
| AK292847.1 | - | 10 | 993 | |||
| AK310863.1 | - | 10 | 993 | |||
| AL133600.1 | - | 10 | 1018 | |||
| BC028740.2 | - | 10 | 1256 | 5260998 | ||