Search
Search

TESK2| Gene Symbol: TESK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320800.1 | NP_001307729.1 | 10 | 1552 | ||
| NM_007170.2 | NP_009101.2 | 11 | 1860 | |||
| XM_005270355.3 | XP_005270412.1 | 10 | 1685 | |||
| XM_006710287.2 | XP_006710350.1 | 12 | 1900 | |||
| XM_011540497.1 | XP_011538799.1 | 13 | 1933 | |||
| XM_017000047.1 | XP_016855536.1 | 11 | 1918 | |||
| XM_017000048.1 | XP_016855537.1 | 12 | 2188 | |||
| XM_017000049.1 | XP_016855538.1 | 10 | 1985 | |||
| GenBank mRNA | AB057597.1 | - | 11 | 1852 | ||
| AJ132545.1 | - | 10 | 1804 | |||
| AK027573.1 | - | 11 | 1834 | |||
| AK294172.1 | - | 10 | 1527 | |||
| BC022194.1 | - | 11 | 1840 | |||
| BC033085.1 | - | 9 | 1384 | 5425494 | ||
| BX648441.1 | - | 5 | 1018 | |||