Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007038.4 | NP_008969.2 | 8 | 3508 | ||
XM_011529448.2 | XP_011527750.1 | 7 | 3340 | |||
GenBank mRNA | AF086299.1 | - | 1 | 53 | ||
AF141293.1 | - | 6 | 1543 | |||
AF142099.1 | - | 8 | 2901 | |||
BC093775.1 | - | 8 | 2863 | 7939620 | ||
BC093777.1 | - | 8 | 2863 | 7939622 |