Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HRH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007232.2 | NP_009163.2 | 3 | 1314 | ||
XM_005260266.3 | XP_005260323.1 | 3 | 1017 | |||
XM_017027623.1 | XP_016883112.1 | 3 | 893 | |||
GenBank mRNA | AB019000.1 | - | 3 | 1017 | ||
AF140538.1 | - | 3 | 1315 | |||
AF321910.1 | - | 3 | 1017 | |||
BC096840.1 | - | 3 | 1033 | 6971899 |
Set Membership: |
G-protein coupled receptor |