Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SP140 SIRNA Assays ›
Gene Symbol:
SP140
Gene Name:
SP140 nuclear body protein
Gene Aliases:
LYSP100, LYSP100-A, LYSP100-B
Chromosome Location:
Chr.2: 230202924 - 230316571 on Build GRCh38
UniGene ID:
Hs.632549
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS117513
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SP140
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005176.2
NP_001005176.1
3
481
NM_001278451.1
NP_001265380.1
3
481
NM_001278452.1
NP_001265381.1
3
481
NM_001278453.1
NP_001265382.1
3
481
NM_007237.4
NP_009168.4
3
481
XM_005246252.2
XP_005246309.1
3
445
XM_005246253.2
XP_005246310.1
3
445
XM_005246254.2
XP_005246311.1
3
445
XM_005246255.2
XP_005246312.1
3
445
XM_005246256.2
XP_005246313.1
3
445
XM_006712223.3
XP_006712286.1
3
445
XM_011510515.1
XP_011508817.1
3
445
XM_011510516.1
XP_011508818.1
3
445
XM_011510517.2
XP_011508819.1
5
1309
XM_011510518.2
XP_011508820.1
4
581
XM_011510519.1
XP_011508821.1
3
445
XM_011510520.1
XP_011508822.1
3
445
XM_017003239.1
XP_016858728.1
3
445
XM_017003240.1
XP_016858729.1
3
445
XM_017003241.1
XP_016858730.1
3
445
XM_017003242.1
XP_016858731.1
3
445
XM_017003243.1
XP_016858732.1
3
445
XM_017003244.1
XP_016858733.1
4
581
XM_017003245.1
XP_016858734.1
3
445
XM_017003246.1
XP_016858735.1
3
445
XM_017003247.1
XP_016858736.1
3
445
XM_017003248.1
XP_016858737.1
3
445
XM_017003249.1
XP_016858738.1
3
445
XM_017003250.1
XP_016858739.1
3
445
XM_017003251.1
XP_016858740.1
4
588
XM_017003252.1
XP_016858741.1
3
445
GenBank mRNA
AA741304.1
-
2
370
AK301267.1
-
3
481
AK301317.1
-
3
482
BC038091.1
-
3
452
BC054890.1
-
3
451
BC070160.1
-
3
447
BC105743.1
-
3
464
40035747
BC105744.1
-
3
466
40035746
BC105960.1
-
3
466
40035752
DC391925.1
-
3
481
U36499.1
-
3
482
U36500.1
-
3
482
U63420.1
-
3
472
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
defense response
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0