Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PALLD SIRNA Assays ›
Gene Symbol:
PALLD
Gene Name:
palladin, cytoskeletal associated protein
Gene Aliases:
CGI-151, CGI151, MYN, PNCA1, SIH002
Chromosome Location:
Chr.4: 168497039 - 168928457 on Build GRCh38
UniGene ID:
Hs.151220
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118046
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PALLD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166108.1
NP_001159580.1
18, 19
3260
NM_001166109.1
NP_001159581.1
16, 17
2374
NM_001166110.1
NP_001159582.1
9, 10
1873
NM_016081.3
NP_057165.3
17, 18
3209
XM_005262861.4
XP_005262918.1
19, 20
3959
XM_005262866.2
XP_005262923.1
18, 19
3509
XM_011531768.2
XP_011530070.1
20, 21
4213
XM_011531769.2
XP_011530071.1
19, 20
4162
XM_011531770.2
XP_011530072.1
20, 21
4213
XM_011531771.2
XP_011530073.1
18, 19
3940
XM_011531772.2
XP_011530074.1
17, 18
3841
XM_011531773.1
XP_011530075.1
19, 20
3541
XM_011531774.1
XP_011530076.1
18, 19
3490
XM_011531775.1
XP_011530077.1
18, 19
2894
XM_017007910.1
XP_016863399.1
19, 20
4162
GenBank mRNA
AB023209.1
-
9, 10
1837
AF077041.1
-
5, 6
1323
AF151909.1
-
5, 6
1229
AF464873.1
-
17, 18
3209
AK000845.1
-
5, 6
798
AK025843.1
-
5, 6
800
AK095512.1
-
16, 17
2336
BC013867.2
-
8, 9
1053
BC140850.1
-
17, 18
3171
9021718
BC144666.1
-
18, 19
3222
9053197
BX537391.1
-
1, 2
111
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008771231, XM_008771233, XM_008771232.
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
keratinocyte development
epithelial cell morphogenesis
cytoskeleton organization
cell migration
actin cytoskeleton organization
actin binding
protein binding
muscle alpha-actinin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0