Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WWC1 SIRNA Assays ›
          Gene Symbol:
          WWC1
          Gene Name:
          WW and C2 domain containing 1
          Gene Aliases:
          HBEBP3, HBEBP36, KIBRA, MEMRYQTL, PPP1R168
          Chromosome Location:
          Chr.5: 168292060 - 168472303 on Build GRCh38
          UniGene ID:
          Hs.484047
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS118522
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WWC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001161661.1 NP_001155133.1 9, 10 1281
          NM_001161662.1 NP_001155134.1 9, 10 1281
          NM_015238.2 NP_056053.1 9, 10 1281
          XM_005265850.1 XP_005265907.1 9, 10 1238
          XM_005265853.2 XP_005265910.1 9, 10 1238
          XM_011534485.1 XP_011532787.1 11, 12 1421
          XM_011534486.1 XP_011532788.1 11, 12 1421
          XM_011534487.1 XP_011532789.1 11, 12 1421
          XM_011534488.1 XP_011532790.1 11, 12 1421
          XM_011534489.1 XP_011532791.1 11, 12 1421
          XM_011534490.1 XP_011532792.1 11, 12 1421
          XM_011534491.1 XP_011532793.1 11, 12 1421
          XM_017009276.1 XP_016864765.1 11, 12 1421
          XM_017009277.1 XP_016864766.1 11, 12 1784
          XM_017009278.1 XP_016864767.1 9, 10 1673
          GenBank mRNA AB020676.1 - 4, 5 517
          AF506799.1 - 9, 10 1254
          AF530058.1 - 4, 5 585
          AK126376.1 - 9, 10 1254
          AK296323.1 - 9, 10 1191
          BX640827.1 - 9, 10 1085

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          cell migration
          establishment of cell polarity
          regulation of intracellular transport
          hippo signaling
          regulation of hippo signaling
          negative regulation of hippo signaling
          positive regulation of MAPK cascade
          negative regulation of organ growth
          transcription coactivator activity
          protein binding
          kinase binding
          protein binding, bridging
          protein complex scaffold

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline