Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM1 SIRNA Assays ›
Gene Symbol:
R3HDM1
Gene Name:
R3H domain containing 1
Gene Aliases:
R3HDM
Chromosome Location:
Chr.2: 135531429 - 135725270 on Build GRCh38
UniGene ID:
Hs.412462
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118877
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282798.1
NP_001269727.1
21
2877
NM_001282799.1
NP_001269728.1
18
2485
NM_001282800.1
NP_001269729.1
18
2534
NM_015361.3
NP_056176.2
21
2869
XM_005263626.3
XP_005263683.1
21
2675
XM_005263627.3
XP_005263684.1
21
2672
XM_005263628.3
XP_005263685.1
20
2606
XM_005263629.3
XP_005263686.1
20
2603
XM_006712389.2
XP_006712452.1
21
2861
XM_011510870.1
XP_011509172.1
22
2963
XM_011510871.1
XP_011509173.1
22
2979
XM_011510872.2
XP_011509174.1
21
2921
XM_011510873.2
XP_011509175.1
22
2915
XM_011510874.2
XP_011509176.1
20
2831
XM_011510875.2
XP_011509177.1
20
2847
XM_011510876.2
XP_011509178.1
22
2777
XM_011510877.2
XP_011509179.1
22
2729
XM_011510878.2
XP_011509180.1
21
2708
XM_011510879.2
XP_011509181.1
21
2660
XM_011510881.1
XP_011509183.1
22
2963
XM_017003718.1
XP_016859207.1
21
2874
XM_017003719.1
XP_016859208.1
20
2853
XM_017003720.1
XP_016859209.1
21
2826
XM_017003721.1
XP_016859210.1
22
2793
XM_017003722.1
XP_016859211.1
19
2729
XM_017003723.1
XP_016859212.1
19
2745
XM_017003724.1
XP_016859213.1
19
2564
XM_017003725.1
XP_016859214.1
23
3055
XM_017003726.1
XP_016859215.1
23
3125
XM_017003727.1
XP_016859216.1
22
3083
XM_017003728.1
XP_016859217.1
20
2558
XM_017003729.1
XP_016859218.1
21
2900
XM_017003730.1
XP_016859219.1
19
2644
XM_017003731.1
XP_016859220.1
22
2974
XM_017003732.1
XP_016859221.1
22
2949
XM_017003733.1
XP_016859222.1
23
3098
XM_017003734.1
XP_016859223.1
24
3258
XM_017003735.1
XP_016859224.1
21
2856
XM_017003736.1
XP_016859225.1
20
2686
GenBank mRNA
AK026987.1
-
3
360
AK127793.1
-
18
2505
AK290015.1
-
21
2763
AK297000.1
-
13
1655
BC041093.1
-
21
2811
5556247
D21852.1
-
18
2439
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-xjq9k:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0