Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: WBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012478.3 | NP_036610.2 | 5 | 549 | ||
GenBank mRNA | AK057881.1 | - | 5 | 454 | ||
AK128407.1 | - | 3 | 1679 | |||
AK293214.1 | - | 6 | 603 | |||
AK300875.1 | - | 4 | 371 | |||
BC007452.1 | - | 5 | 452 | 3140484 | ||
BC010616.1 | - | 5 | 462 | 4177319 |