Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DCAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015397.3 | NP_056212.1 | 8 | 1471 | ||
XM_011517843.1 | XP_011516145.1 | 8 | 1298 | |||
GenBank mRNA | AB067479.1 | - | 8 | 1391 | ||
AF131771.1 | - | 3 | 331 | |||
AK000354.1 | - | 8 | 1249 | |||
AK292935.1 | - | 8 | 1277 | |||
AL117646.1 | - | 5 | 605 | |||
BC008893.2 | - | 7 | 1034 | |||
BC063823.1 | - | 8 | 1389 | 6184796 | ||
DQ518909.1 | - | 8 | 1391 |