Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GAPVD1 SIRNA Assays ›
          Gene Symbol:
          GAPVD1
          Gene Name:
          GTPase activating protein and VPS9 domains 1
          Gene Aliases:
          GAPEX5, GAPex-5, RAP6
          Chromosome Location:
          Chr.9: 125261794 - 125365011 on Build GRCh38
          UniGene ID:
          Hs.495134
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS119838
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GAPVD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282679.1 NP_001269608.1 17, 18 3246
          NM_001282680.1 NP_001269609.1 17, 18 3246
          NM_001282681.1 NP_001269610.1 14, 15 2851
          NM_015635.3 NP_056450.2 16, 17 3161
          XM_005251901.3 XP_005251958.1 16, 17 3110
          XM_005251904.3 XP_005251961.1 16, 17 3107
          XM_006717044.3 XP_006717107.1 16, 17 3032
          XM_011518499.2 XP_011516801.1 17, 18 3205
          XM_011518500.2 XP_011516802.1 18, 19 3322
          XM_011518502.2 XP_011516804.1 17, 18 3133
          XM_011518506.2 XP_011516808.1 17, 18 3130
          XM_011518507.2 XP_011516809.1 17, 18 3210
          XM_017014600.1 XP_016870089.1 16, 17 3015
          XM_017014601.1 XP_016870090.1 15, 16 3037
          XM_017014602.1 XP_016870091.1 17, 18 3257
          XM_017014603.1 XP_016870092.1 17, 18 3204
          XM_017014604.1 XP_016870093.1 18, 19 3306
          XM_017014605.1 XP_016870094.1 16, 17 3118
          XM_017014606.1 XP_016870095.1 17, 18 3151
          XM_017014607.1 XP_016870096.1 17, 18 3160
          XM_017014608.1 XP_016870097.1 15, 16 2970
          XM_017014609.1 XP_016870098.1 17, 18 3219
          XM_017014610.1 XP_016870099.1 15, 16 3037
          XM_017014611.1 XP_016870100.1 16, 17 3030
          GenBank mRNA AB040954.3 - 17, 18 3172
          AK000126.1 - 12, 13 2392
          AK023841.1 - 8, 9 1195
          AK027049.1 - 13, 14 1932
          AK056493.1 - 16, 17 3123
          AK294542.1 - 15, 16 3037
          AK296381.1 - 16, 17 3170
          AL080196.1 - 6, 7 943
          BC010027.2 - 15, 16 3000
          BC013635.1 - 1, 2 232
          BC021119.1 - 13, 14 2570
          BC114937.1 - 17, 18 3129 40083103
          BC114962.1 - 15, 16 2965 40083104
          DQ233254.1 - 14, 15 2819

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          signal transduction
          positive regulation of GTPase activity
          regulation of protein transport
          cell-cell adhesion
          guanyl-nucleotide exchange factor activity
          GTPase activator activity
          GTPase activating protein binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mc8hj:80/100.66.75.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0